Learn More
Enzo Life Sciences DiOC6(3) iodide (ultra pure), (100mg), CAS Number: 53213-82-4
SDP
Supplier:  Enzo Life Sciences ENZ52303
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Mitochondria dye . Alternative Name: 3,3'-Dihexyloxacarbocyanine iodide. MW: 572.5. CAS Number: 53213-82-4. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Use/Stability: Stable for at least one year after receipt when stored as recommended. Long Term Storage: -20°C
Catalog No. NC0664890