Learn More
Enzo Life Sciences Dimethyl-W84 dibromide (5mg). CAS: 402475-33-6
SDP
Supplier:  Enzo Life Sciences ALX550400M005
Encompass_Preferred
Very potent allosteric modulator of muscarinic receptors, selective for M2-receptors. Retards the dissociation of the antagonist N-methylscopolamine. Affinity for the free receptor is about 10-fold higher than with W84 (ALX-550-399). pKA=7.08, p(αKA)=6.75, pEC50,diss=6.77. Alternative name: N,N-Bis[3-(1,3-dihydro-5-methyl-1,3-dioxo-2H-isoindol-2-yl)propyl]-N,N,N,N-tetramethyl-1,6-hexanediamminium dibromide. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO, water or aqueous buffer. Long Term Storage: +4°C.
Catalog No. 50-200-9552
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.