Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl tert-butylmalonate, 95%, Thermo Scientific Chemicals

Catalog No. AAH2653203
Click to view available options
Quantity:
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39520-25-7
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD08436110
InChI Key OBANUQSDPDUGSL-UHFFFAOYSA-N
PubChem CID 13574047
IUPAC Name dimethyl 2-tert-butylpropanedioate
SMILES COC(=O)C(C(=O)OC)C(C)(C)C

Specifications

Refractive Index 1.428
Quantity 1 g
Synonym dimethyl 2-tert-butyl malonate, dimethyl tert-butylmalonate, propanedioic acid, 1,1-dimethylethyl-, dimethyl ester, 1,3-dimethyl 2-tert-butylpropanedioate, acmc-1aijh, dimethyl2-tert-butyl malonate, dimethyl tert-butylpropanedioate, tert-butylmalonic acid dimethyl ester, propanedioic acid,2-1,1-dimethylethyl-, 1,3-dimethyl ester
Formula Weight 188.22
Percent Purity 95%
Chemical Name or Material Dimethyl tert-butylmalonate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.