Learn More
New England Biolabs, Inc. Diluent C with rAlbumin - 5 ml
Supplier: New England Biolabs, Inc. B8534S
Diluent C (with rAlbumin) is recommended for making dilutions of some NEB restriction enzymes. When necessary, we recommend diluting enzymes just prior to use and suggest that the final concentration of diluted enzymes be at least 1,000 units/ml. Restriction enzymes should be stored at their selling concentrations. Each vial contains 1.3ml of diluent buffer.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.