Learn More
New England Biolabs, Inc. Diluent B with rAlbumin - 5 ml
SDP
Supplier:  New England Biolabs, Inc. B8533S
Encompass
Diluent B (with rAlbumin) is recommended for making dilutions of some NEB restriction enzymes. When necessary, we recommend diluting enzymes just prior to use and suggest that the final concentration of diluted enzymes be at least 1,000 units/ml. Restriction enzymes should be stored at their selling concentrations.
Catalog No. 50-252-7905
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.