Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisobutyl Disulfide 98.0+%, TCI America™
SDP

Catalog No. D061625ML
Click to view available options
Quantity:
25 mL

Chemical Identifiers

CAS 1518-72-5
Molecular Formula C8H18S2
Molecular Weight (g/mol) 178.35
MDL Number MFCD00039850
InChI Key ONJROLGQWMBXAP-UHFFFAOYSA-N
Synonym Isobutyl Disulfide
PubChem CID 80284
IUPAC Name 2-methyl-1-[(2-methylpropyl)disulfanyl]propane
SMILES CC(C)CSSCC(C)C

Specifications

Color Yellow
Boiling Point 109°C
Quantity 25 mL
Formula Weight 178.35
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisobutyl Disulfide
EINECSNumber (2)-0477
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.