Learn More
Hach Company Digital Titrator, 0.00125 mL/digit
Supplier:  Hach Company 1690001
Encompass_Preferred
Digital Titrator, 0.00125 mL/digit, with case, manual, and five straight delivery tubes.
Catalog No. 50-199-5159
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.