Learn More
Azzota Corp Digital Stirring Heating Mantle, Capacity: 3000Ml, 350W, Maximum Temperature Is 450'C. Stir Speed: 0 To 1400Rpm.
Supplier:  Azzota Corp DSHM3000
Encompass_Preferred
Digital Stirring Heating Mantle, Capacity: 3000ml, 350W, Maximum temperature is 450'C. Stir speed: 0 to 1400rpm.
Catalog No. 50-161-2830
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.