Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butylphenylphosphine 97.0+%, TCI America™
SDP

Catalog No. D39405G
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 32673-25-9
Molecular Formula C14H23P
Molecular Weight (g/mol) 222.312
MDL Number MFCD00049096
InChI Key XOJNEFQLMRCOMS-UHFFFAOYSA-N
PubChem CID 316378
IUPAC Name ditert-butyl(phenyl)phosphane
SMILES CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C

Specifications

Color Colorless
Boiling Point 128°C
Quantity 5 g
UN Number 2845
Formula Weight 222.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-tert-butylphenylphosphine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.