Learn More
Enzo Life Sciences Di-8-ANEPPS (5mg). CAS: 157134-53-7
Supplier: Enzo Life Sciences ENZ52204
Di-8-ANEPPS is a member of the ANEP class of membrane potential dyes. These dyes are weakly fluorescent in aqueous media, and become strongly fluorescent upon binding to lipophilic environments such as membranes. A change in the surrounding electronic field demonstrates a membrane potential-dependent shift in excitation spectra. Di-8-ANEPPS is less susceptible for cellular internalization than other ANEP dyes probably due to a sulfonate group. The response is sufficiently fast to detect transient (millisecond) potential changes in excitable cells, including single neurons, cardiac cells and intact brains. Wavelength Maxima: Excitation 498nm, Emission 713nm Alternative name: 4-[2-[6-(Dioctylamino)-2-naphthalenyl]ethenyl]-1-(3-sulfopropyl)-pyridinium, inner salt. Purity: ≥90% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.