Learn More
Cole-Parmer Shimpo DT-311A Digital Stroboscope; 115 VAC
Supplier:  Cole-Parmer 9880633
Encompass
The DT-311A stroboscope provides the illusion of "stopped motion." By adjusting the strobe’s flash rate, equipment appears to be standing still or in slow motion. The stroboscope also measures rotational (RPM) or reciprocating (strokes per minute) speeds with the same precision as an electronic digital tachometer. Ideally suited for printing inspection, motion analysis, preventive maintenance, and speed measurement of repetitive motion. The strobes provide timely data on vibrating parts, leaks, spray patterns and a variety of industrial and scientific operations. It is extremely easy to maneuver and operate via its lightweight and compact design and simplified user interface. Its rugged metal construction provides unequaled reliability in the harshest environments.
Catalog No. 50-993-609
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.