Learn More
World Precision Instrument Desmarres Lid Retractor, 14 cm, 13 mm wide Blade
Supplier: World Precision Instrument WP108220
DeMarres Lid Retractors are surgical instruments used in ophthalmic surgery to hold back the eyelids and expose the eye for examination or surgical procedures. The DeMarres lid retractor is designed to provide a clear and unobstructed view of the eye, particularly during surgeries involving the anterior segment of the eye. The blade of the rectractor is curved to follow the contour of the eye, giving optimal exposure with limited discomfort. The tip of the blade is rounded and blunt to avoid injury to delicate eye tissue. Choose the size of the retractor you need: 0 (11 mm wide), 1 (13 mm wide), and 2 (15 mm wide). The square handles are lightly textured and polished for a solid grip. These retractors are made of surgical grade stainless steel and are suitable for autoclaving.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.