Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Public Seating™ Designer Science Table
SCI_ED

High end designer adjustable legs for science and art environments

Supplier:  National Public Seating™ SLT62460H

 View more versions of this product

Catalog No. S83713


Explore all special offers
Add to Cart

Description

Description

  • Under table hat support for 500 lb. weight capacity
  • Tables adjust in 3/4 in. increments from 27 to 42 in. Height
  • 11-gauge leg-bases are fastened to the tabletop with 6 heavy-duty screws
  • All legs come standard with simple-adjust glides
  • 10 Year Warranty
Specifications

Specifications

Table
HPL
Art and Science Environments
10 years
500 lb.
MAS2
60 x 24 x 27 to 42 in.
Black legs
Black
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.