Learn More
Electron Microscopy Sciences Desiccant in A Cartridge 160 mm
Supplier:  Electron Microscopy Sciences 7120916
Encompass_Preferred
The desiccant in a cartridge regenerates repeatedly without degradation. These cartridges fit into the bottom of a standard glass or plastic desiccator. The cartridge consists of a durable long wearing polyester felt bag with a perforated Mylar cover sealed in place. The indicating gel beads will change their color from blue when dry to pink when moist. Saturated gel can be regenerated in a 150 degree C (300 degree F) vented oven for 3 hours or more (depending on size), until the blue color is restored.
Cartridges come individually sealed in a moisture-proof pouch ready to use.
Catalog No. 50-930-893
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.