Learn More
Electron Microscopy Sciences Desiccant Cartridge, Large
Supplier:  Electron Microscopy Sciences 7121204
Encompass_Preferred
These desiccant cartridges can be used in desiccator cabinets or non-vacuum desiccators, analytical balance cases, and precision laboratory instruments. The clear polystyrene dish allows for instant visual inspection of the silica gel color which changes from blue to pale pink as it absorbs moisture.
A highly porous membrane covers the dish of the desiccant providing good moisture absorption. The desiccant can be regenerated by heating cartridges in an oven at 50 degree C (122 degree F) overnight or in a 150 degree C (302 degree F) oven for one hour. The silica gel is inert and non-toxic. Each cartridge is individually sealed in aluminum foil. Dispose of the cartridges after repeated use.
Catalog No. 50-930-899
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.