Learn More
Electron Microscopy Sciences DER 736 Resin 225 ML
Supplier: Electron Microscopy Sciences 13001
A light colored, flexible epoxy. Used to simplify infiltration in combination with Epon 812 (Embed 812). Kushida, J. Electron Micro., 16,278 (1964)