Learn More
Electron Microscopy Sciences DER 332 Resin 225 ML
Supplier:  Electron Microscopy Sciences 13030
Encompass_Preferred
A relatively soft embedding resin used when slower polymerization is needed. Lockwood, Anat. Rec., 150,129 (1964).
Catalog No. 50-980-345
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.