Learn More
Hach Company Demineralizer Bottle, 473 mL Capacity
Supplier:  Hach Company 2184600
Encompass_Preferred
Narrow mouth, polyethylene bottle. Contains monobed resin. Flip spout. Great way to deionize small volumes of water. Used with Hach Portable Laboratories.
Catalog No. 50-199-5526
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.