Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fischer Technical Company™ Dehydrated Culture Media and Ingredients
Essential agents for preparing culture media.
Supplier: Fischer Technical Company™ CD150A
Description
Successful bacteriological work begins with high-quality media.
- Agar, a solidifying agent, is an essential component of many media
- Available in a variety of formulations and bottle sizes
- For use in preparing plated media, tubed media, bottled media, and slants
Specifications
Dermatophytes and Fungi | |
Bottled | |
Sabouraud Dextrose Agar | |
Non-sterile | |
Microbiological Tests | |
113 g |
Dehydrated Powder | |
US with Globally Sourced Materials | |
Microbiology Media | |
Fungi | |
Microbiological Testing |
Safety and Handling
ShelfLife : 5 Years
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction