Learn More
New England Biolabs, Inc. Deep Vent™ DNA Polymerase – 200 units
Supplier: New England Biolabs, Inc. M0258S
Deep Vent DNA Polymerase was the second high-fidelity thermophilic DNA polymerase available from New England Biolabs. The fidelity of Deep Vent DNA Polymerase is derived in part from an integral 3 5 proofreading exonuclease activity. Deep Vent is even more stable than Vent at temperatures of 95 to 100C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.