Learn More
Andwin Scientific DECONQUAT 2:128/1GAL STRL 4CS
Supplier: Andwin Scientific 14003702
DECON-QUAT 100 Quaternary Ammonium Solution Broad spectrum hospital disinfectant fungicide deodorizer and more. Effective against Hepatitis B Virus. Filtered at 0.2u. SimpleMix system 2:128 in 3.8L of WFI water. Gamma irradiated.