Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ DDR1-IN-1 dihydrochloride
Selective DDR1 inhibitor
$342.50 - $1396.00
Specifications
Purity | >98% |
---|---|
CAS | 1780303-76-5 |
Product Type | DDR1-IN-1 dihydrochloride |
Molecular Weight (g/mol) | 625.51 |
Inhibitors | DDR1-IN-1 dihydrochloride |
Description
Selective discoidin domain receptor 1 (DDR1) tyrosine kinase inhibitor (IC50 values are 105 and 413 nM for DDR1 and DDR2 respectively). Inhibits integrin-induced DDR1 autophosphorylation in an osteosarcoma cell line.Specifications
>98% | |
DDR1-IN-1 dihydrochloride | |
DDR1-IN-1 dihydrochloride |
1780303-76-5 | |
625.51 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title