Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ DDIT3/FUS SY Translocation FISH Probe
Labeled FISH probes for identification of gene transloaction using Fluoresecent In Situ Hybridization Technique.
$3710.00
Specifications
Content And Storage | Store at 4°C in the dark. |
---|---|
Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Format | Liquid |
For Use With (Application) | FISH-Ce |
Gene ID (Entrez) | 1649/2521 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-941-600
|
Abnova™
FT0018 |
100 μL |
Each of 1 for $3,710.00
|
|
|||||
Specifications
Store at 4°C in the dark. | |
Liquid | |
1649/2521 |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Spot an opportunity for improvement?
Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title