Learn More
Andwin Scientific CYTOSEAL XY1 MOUNT MEDIUM 4OZ
Supplier: Andwin Scientific M76311
The Cytoseal™ XYL Mounting Medium is composed of high-quality synthetic resins and antioxidant that eliminates the chance of fading or yellowing of specimens. This product can also be dried quickly, which permits specimen examinations right after the application of the medium. It is packaged and sold in a 4 oz. bottle that has a twist-open cap.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.