Learn More
Enzo Life Sciences CYTO-ID Autophagy detection kit (1x200tests)
Supplier: Enzo Life Sciences ENZ51031K200
A no-transfection, quantitative assay for monitoring autophagy in live cells. CYTO-ID.The probe is a cationic amphiphilic tracer (CAT) dye that rapidly partitions into cells in a similar manner as drugs that induce phospholipidosis. Careful selection of titratable functional moieties on the dye prevents its accumulation within lysosomes, but enables labeling of vacuoles associated with the autophagy pathway. Applications: Flow Cytometry. Fluorescence microscopy. Fluorescent detection, HTS. Contents: CYTO-ID Green Detection Reagent. Hoechst 33342 Nuclear Stain. Autophagy Inducer (Rapamycin). Chloroquine Control. 10X Assay Buffer
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.