Learn More
ACROBiosystems Human C-Reactive Protein (CRP) ELISA Kit
Supplier:  ACROBiosystems CRSB01696TESTS
Encompass_Preferred
ClinMax™ ELISA Kit is convenient ready-to-use immunoassay Kit, specifically designed to quantitate human CRP that is present in complex biological samples, such as human serum, plasma, and cell culture supernates. A comprehensive validation of the ELISA method was performed following the ICH M10 on bioanalytical method validation and the FDA’s bioanalytical method validation guidance for industry. This validation included assessments of linearity, accuracy, precision, dilution linearity, recovery, and the hook effect. For details information, please refer to the DS. ClinMax™ ELISA Kits are manufactured in a GMP-like facility and comply to the ISO 13485 standard, ensuring a high level of quality and reliability.
Catalog No. 50-271-9689
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.