Learn More
Enzo Life Sciences Cylindrospermopsin (1mg). CAS: 143545-90-8
Supplier: Enzo Life Sciences ALX350149M001
Tricyclic alkaloid hepatotoxin. Exhibits a completely different mechanism of toxicity than microcystins. Protein synthesis inhibitor. Might be carcinogenic. Inhibits pyrimidine nucleotide synthesis. Purity: ≥95% (HPLC). Appearance: Glassy solid. Solubility: Soluble in water, methanol or DMSO. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.