Learn More
Sklar Instruments Forceps, Cushing Dressing Forceps, Sklar 7 in., Cushing Dressing Forceps Gutch Handle, Cross-Serrated, Tungsten Carbide / Stainless Steel, EA.
Supplier: Sklar Instruments 514398
Forceps, Cushing Dressing Forceps, Sklar, Cushing Dressing Forceps Gutch Handle, Cross-Serrated, Tungsten Carbide / Stainless Steel, EA. Sklar Tungsten Carbide Cushing Dressing Forceps have thin, long tips that are ideal for reaching narrow spaces. They are available with different handle types to accommodate various procedures and user preference. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles. This product is straight with cross-serrated tips and a length of 7 inches. It features a gutch handle. Lifetime warranty against all manufacturer defects.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.