Learn More
Andwin Scientific CULTURE TUBES, DISP BS 13X100
SDP
Supplier:  Andwin Scientific 47729572
Encompass_Preferred
The Culture Tubes feature resistance against chemical attack and weathering. In order to remove contamination, increase its strength, and lessen the chance of breakage, each tube is heat-treated at temperatures exceeding 500C (932F) immediately bef
Catalog No. NC0779746
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.