Learn More
Thermo Scientific™ Culti-Loops™ Citrobacter diversus KM 11012
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Supplier: Thermo Scientific™ R4601508
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Citrobacter species | |
Citrobacter diversus KM 11012 | |
Gram-negative Rods | |
5 Loops/Pack |
KM 11012TM* | |
Culti-Loops Quality Control Organism | |
Qualitative QC |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.