Learn More
Hampton Research CRYSTALQUICK 96 WELL PL 20CSC
Supplier: Hampton Research HR3284
HR3-284/CrystalQuick 96 Well Sitting Drop Plate (Greiner 609871), 4 ul square drop well, 1 drop well, LBR, low profile - 20 plate case. Please note items from this supplier are non-returnable.