Learn More
Hampton Research CRYSTALQUICK 96 WELL PL 10CSC
SDP
Supplier:  Hampton Research HR3192
Encompass
HR3-192/CrystalQuick 96 Well Sitting Drop Plate (Greiner 609101), 4 ul square drop well, 3 drop well - 10 plate case. Please note items from this supplier are non-returnable.
Catalog No. 50-018-6
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.