Learn More
Andwin Scientific CRYSTAL/EPPEN TIP 10 RACKS/CS
SDP
Supplier:  Andwin Scientific 7244202
Encompass_Preferred
Specimen Preparation and Embedding Supplies: Pipettes and Pipette Fillers: Pipette TipsKENDALLTreated Pipette Tips: 0.5 -10ul Crystal/Eppendorf Tips-Natural, Non-Sterile, 96Tips /Rack, 10 Racks/case
Catalog No. 50-949-401
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.