Learn More
Sarstedt Inc Tube, Cryogenic, 2.0mL, Internal Thread, Green Cap, 500/Pack
Supplier: Sarstedt Inc 72.380.005
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.6mL; With internal thread; Sterile; Green cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.