Learn More
Sarstedt Inc Tube, Cyrogenic, 2.0mL, External Thread, White Cap, 500/Pack
Supplier:  Sarstedt Inc 72.379
Encompass_Preferred
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.8mL; With external thread; Sterile; White cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.
Catalog No. 50-873-794
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.