Learn More
Sarstedt Inc Tube, Cyrogenic, 2.0mL, External Thread, Yellow Cap, 500/Pack
Supplier: Sarstedt Inc 72.379.004
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.8mL; With external thread; Sterile; Yellow cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.