Learn More
Sarstedt Inc Tube, Cryogenic, 1.2mL, External Thread, White Cap, 500/Pack
Supplier: Sarstedt Inc 72.377
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.0mL; With external thread; Sterile; White cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.