Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ad Infinitum™ CryoProtectPURE™ Cryopreservation Media PROMO
Ad Infinitum™ CryoProtectPURE™ Cryopreservation Media is a DMSO-free and Serum-free cryopreservation media for cell and tissue preservation.
Supplier: Ad Infinitum™ CPP100025PM
Description
- Increases the cell viability during freeze and post-thaw, producing healthier cells with no antigenicity concern or toxic vitrification.
- Serum-free
- Pre-mixed
- Ready-to-use
- Filtered to 0.2 μm
- Suitable for a wide variety of cells
- Shipped at ambient temperature
- For manufacturing and/or research use only, not for diagnostic or therapeutic use
- Manufactured at a cGMP-compliant facility registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards
Specifications
ISO 13485 standards | |
Pre-mixed and ready-to-use | |
Manufacturing and/or Research Use Only, Not for Diagnostic or Therapeutic Use |
2°C to 8°C | |
Cryopreservation Media | |
25 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction