Learn More
Andwin Scientific Direct thermal Cryo-Tags 1.50x0.50inch, Orange, 750/roll
Supplier: Andwin Scientific DTCR2000O
Direct Thermal Cryo-Tags are formatted to easily work with the Dymo Labelwriter, but also with most direct thermal printers. Quickly label your samples with sharp text, clear barcodes or a combination of both. Temperatures From 70 to -196degreeC.