Learn More
MTC Bio Cryogenic Vials, internally threaded caps with silicone O-ring, 3mL, sterile, printed graduations and marking area, self-standing star foot, 50-place locking rack included with each case, 10 resealable bags of 50 tubes, 500/cs
Supplier:  MTC Bio V4803
Encompass_Preferred
MTC Bio cryogenic vials are designed to hold, store and preserve biological materials for days, weeks, years, or decades. These thick-walled tubes are outfitted with silicone O-rings that allow them to maintain their seal at temperatures as low as -196°C. Sterilized by gamma irradiation, the tubes are packed in reasealable bags of 50 tubes (10 bags per case) and are sterilzied by gamma. The self-standing starfoot base mates perfectly with a FREE included rack so that one hand is all that is need to install and remove caps. RNase, DNase, pyrogen, & endotoxin free
Catalog No. 50-226-8512
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.