Learn More
Andwin Scientific CRYO3 CryoMold, Standard, 100/cs
SDP
Supplier:  Andwin Scientific 6253201
Encompass_Preferred
Plastic Cryomold, round shapes with a tab for easy handling. Available in standard and biopsy configurations. The standard mold measures 22 mm (D) x 5 mm deep. The Biopsy mold measures 15 mm (D) x 5 mm deep.
Catalog No. NC9614277
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.