Learn More
Chemglass Life Sciences Aluminum Seal, 11mm, Silver, PTFE/Silicone/PTFE
SDP
Supplier:  Chemglass Life Sciences CV34000011
Encompass_Preferred
Aluminum Seal, 11mm, Silver, PTFE/Silicone/PTFE. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. The seal with the viton septa is resistant to many aggressive solvents. The 11mm magnetic seal can be used with any instrument where the vials is transported with the use of a magnet.
Catalog No. 50-230-970
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.