Learn More
Advanced ImmunoChemical, Inc. Canine Parvovirus Vp2 Protein, Recombinant, 0.1mg
Supplier:  Advanced ImmunoChemical, Inc. 8CPV20.1MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Antigens / Veterinary / Recombinant canine parvovirus VP2. Expressed in an insect cell lineDescription:Recombinant canine parvovirus variant 2b (CPV-2b) capsid protein VP2 consists of 584 amino acid residues.Theoretical molecular weight 64657 Da.Theoretical pI 5.36.Does not contain any affinity tags.Applications:Calibrator or standard in immunoassays for CPV immunodetection or antigen in immunoassays for determination of CPV antibody titre in canine blood.Analysis:Purity > 90% (SDS-PAGE in reducing conditions)Concentration is determined spectrophotometrically.Immunoreactivity is confirmed by reaction with monoclonal antibodies (Cat. #3PV16) specific to canine parvovirus.Presentation:Lyophilized from 20 mM Na-phosphate, pH 7.4, 30 mM NaCl, 0.01% Tween 20, 10% trehalose.It is recommended to reconstitute this product with deionized water to its initial concentration.
Catalog No. 50-153-9839
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.