Learn More
Innovative Research Inc Cow Plsmngn Aff Pur 1mg
Supplier:  Innovative Research Inc IBOPLGAP1MG
Encompass_Preferred
Bovine Plasminogen Affinity Purified 1mg - Bovine Plasminogen Affinity Purified from Innovative Research has been purified from bovine plasma by affinity chromatography. This is a Frozen liquid buffered in 0.1M HEPES; 0.1M NaCl; pH 7.4 and a purity of 95% as determined by SDS-PAGE. Additional Information Prepared from bovine plasma by immobilized lysine chromatography. This product can be used as a control in applications like ELISA and Western Blot. It can be used in a wide range of in vitro applications, including cell differentiation, cell signaling, clotting assays, fibrinolysis, plasminogen activation, protein-protein interactions, receptor binding, thromboelastography, thrombolysis, zymography, and more.
Catalog No. 50-274-4622
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.