Learn More
Andwin Scientific COVERGLASS #1 24X60 EA
SDP
Supplier:  Andwin Scientific 6377001
Encompass_Preferred
The Gold Seal 24 x 60mm with 0.13-0.17mm thickness Cover Glass is made from preselected and precleaned silicate glass. This product comes in a lint free box that can be converted to slide storage. This product is sold an ounce a piece.
Catalog No. 50-727-484
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.