Learn More
Andwin Scientific COVERGLASS #1 22X22 10/PK
Supplier: Andwin Scientific 6375710
The Gold Seal 22X22mm with 0.13-0.17mm thickness Cover Glass is constructed from preselected and precleaned silicate glass. It comes in a lint free box that could be converted to slide storage. It is sold in 10 a pack, ounce each.