Learn More
Corning™ Costar™ Stripettor™ Pipet Filler/Dispenser Replacement Parts
For use with Stripettor™ filler/dispenser
Specifications
For Use With (Equipment) Costar™ stripettor pipet filler/dispenser
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
07-201-792
View Documents
Corning™
4095
Replacement Filter, 0.45μm N/A N/A N/A
07-201-791
View Documents
Corning™
4094
Replacement Filter, 0.2μm N/A N/A N/A
Description
Description
  • PTFE filter (pack of 5) is for the Stripettor™ Plus pipetting controller
  • Pore Size: 0.45 μm
  • Autoclavable
Specifications
Specifications
Costar™ stripettor pipet filler/dispenser
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.