Learn More
Julabo USA Inc CORIO CP immersion heating circulator; +20…+200 °C operating range; ±0.02 °C stability; 2 kW heater, 8-27 L/min pump with 1.5-10.2 psi; adjustable clamp; applications to 50L; USB & RS232 communication
SDP
Supplier:  Julabo USA Inc 9013000.33
Encompass_Preferred
CORIO™ CP, The new generation of laboratory circulators from JULABO.CORIO™ CP immersion heating circulators are suitable for temperature applications up to 50L. Adjustable clamp makes installation easy. Operating range from +20…+200 °C.Bright LED display with easy to use interface. USB and RS232 connections for external PC control and data-logging; PID temperature control provides ±0.02 °C stability. Adjustable centrifugal pump capacity of 8-27 L/min with pressure from 1.5-10.2 psi.Class III (FL) according to DIN 12876-1.
Catalog No. 50-190-4648
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.