Learn More
Sino Biological COPZ1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80636R07E100
Encompass_Preferred
A DNA sequence encoding the Human / Mouse / Rat COPZ1 (NP_001101587.1) (Met1-Arg177) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-2601
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.