Learn More
Glass Expansion Copper Skimmer Cone s-lens
Supplier: Glass Expansion AT8902SCU
Copper Skimmer Cone for s-lens (G3666-67067). For Agilent 8900 ICP-MS. Recommended for semiconductor applications. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For routine applications, use standard Nickel Skimmer for x-lens, see P/N 501907593.For organic analysis, use Platinum Skimmer with Ni Base for x-lens, P/N 501907595, which must be mounted on brass adaptor skimmer base, see P/N AT7905X-BR. For lowest nickel background and laser ablation applications, use Aluminum Skimmer for x-lens, see P/N AT7902X-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.